Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfml3
Entrez Gene ID
310743
Gene Name
olfactomedin-like 3
Gene Aliases
-
Chromosome Location
Chr.2: 206219419 - 206222248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107708.1 NP_001101178.1 2 - 3 481 73
XM_006233077.1 XP_006233139.1 2 - 3 466 73
GenBank mRNA BC158838.1 - 2 - 3 420 73

View Full Product Details