Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acp6
Entrez Gene ID
295305
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
-
Chromosome Location
Chr.2: 199284177 - 199305793 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031645.1 NP_001026815.1 8 - 9 950 82
GenBank mRNA BC099131.1 - 8 - 9 950 82

View Full Product Details