Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Celf3
Entrez Gene ID
499669
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
RGD1563168, Tnrc4
Chromosome Location
Chr.2: 195716360 - 195732872 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232911.2 XP_006232973.1 2 - 3 168 76

View Full Product Details