The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106451.1 | NP_001099921.1 | 3 - 4 | 487 | 69 |
XM_006233027.2 | XP_006233089.1 | 4 - 5 | 1062 | 69 | |
XM_008761285.1 | XP_008759507.1 | 4 - 5 | 5361 | 69 |