The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024303.1 | NP_001019474.1 | 6 - 7 | 847 | 89 |
XM_006232978.2 | XP_006233040.1 | 5 - 6 | 867 | 89 | |
GenBank mRNA | BC087077.1 | - | 6 - 7 | 847 | 89 |