Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx27
Entrez Gene ID
260323
Gene Name
sorting nexin family member 27
Gene Aliases
Mrt1
Chromosome Location
Chr.2: 195738613 - 195821608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110151.1 NP_001103621.1 8 - 9 1350 85
NM_152847.2 NP_690060.2 8 - 9 1350 85
GenBank mRNA AB010245.1 - 8 - 9 1350 85
AB051816.1 - 8 - 9 1350 85

View Full Product Details