Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmgcs2
Entrez Gene ID
24450
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial)
Gene Aliases
Hmgcs1, Mt3h3mg
Chromosome Location
Chr.2: 200452619 - 200480785 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173094.2 NP_775117.2 3 - 4 717 64
XM_006233002.2 XP_006233064.1 3 - 4 746 64
GenBank mRNA BC078695.1 - 3 - 4 717 64
BC083543.1 - 3 - 4 717 64
M33648.1 - 3 - 4 735 64

View Full Product Details