Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclre1b
Entrez Gene ID
310745
Gene Name
DNA cross-link repair 1B
Gene Aliases
-
Chromosome Location
Chr.2: 206285085 - 206293599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025687.1 NP_001020858.1 4 - 5 742 103
XM_006233078.2 XP_006233140.1 3 - 4 824 103
XM_006233079.1 XP_006233141.1 4 - 5 737 103
GenBank mRNA BC098939.1 - 4 - 5 742 103

View Full Product Details