Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpysl2
Entrez Gene ID
25416
Gene Name
dihydropyrimidinase-like 2
Gene Aliases
Crmp2, TOAD-64
Chromosome Location
Chr.15: 43475640 - 43581757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105717.2 NP_001099187.1 2 - 3 350 83
XM_006252097.2 XP_006252159.1 2 - 3 581 83
GenBank mRNA Z46882.1 - 2 - 3 303 83

View Full Product Details