Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo16
Entrez Gene ID
305970
Gene Name
F-box protein 16
Gene Aliases
-
Chromosome Location
Chr.15: 48673727 - 48723370 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013132.2 NP_001013150.1 6 - 7 897 131
XM_006252201.2 XP_006252263.1 7 - 8 1670 131
XM_006252202.2 XP_006252264.1 6 - 7 1550 131
GenBank mRNA BC062801.1 - 6 - 7 900 131
FQ213407.1 - 6 - 7 831 131

View Full Product Details