Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scara5
Entrez Gene ID
305974
Gene Name
scavenger receptor class A, member 5
Gene Aliases
RGD1306539
Chromosome Location
Chr.15: 49069142 - 49177159 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135855.1 NP_001129327.1 3 - 4 578 73
XM_006252204.2 XP_006252266.1 3 - 4 616 73

View Full Product Details