The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025142.2 | NP_001020313.2 | 5 - 6 | 953 | 98 |
GenBank mRNA | BC085942.1 | - | 5 - 6 | 916 | 98 |
BC094955.1 | - | 3 - 4 | 349 | 98 | |
FQ224662.1 | - | 5 - 6 | 946 | 98 |