The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022220.2 | NP_071556.2 | 11 - 12 | 1231 | 114 |
XM_006252146.2 | XP_006252208.1 | 10 - 11 | 1207 | 114 | |
GenBank mRNA | BC089803.1 | - | 11 - 12 | 1231 | 114 |
FQ219140.1 | - | 11 - 12 | 1256 | 114 | |
FQ219632.1 | - | 11 - 12 | 1339 | 114 |