Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shc2
Entrez Gene ID
314612
Gene Name
SHC (Src homology 2 domain containing) transforming protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 12948952 - 12971174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108065.1 NP_001101535.1 11 - 12 1664 106

View Full Product Details