Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Angpt4
Entrez Gene ID
296269
Gene Name
angiopoietin 4
Gene Aliases
Agpt4
Chromosome Location
Chr.3: 147422095 - 147455670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106526.1 NP_001099996.1 7 - 8 1363 80

View Full Product Details