The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057209.1 | NP_476557.1 | 10 - 11 | 1678 | 117 |
GenBank mRNA | J03886.1 | - | 10 - 11 | 1678 | 117 |