Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb29
Entrez Gene ID
641519
Gene Name
defensin beta 29
Gene Aliases
-
Chromosome Location
Chr.3: 148008820 - 148014101 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037368.1 NP_001032445.1 - 111 95
GenBank mRNA AY621360.1 - - 111 95

View Full Product Details