Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map1lc3a
Entrez Gene ID
362245
Gene Name
microtubule-associated protein 1 light chain 3 alpha
Gene Aliases
-
Chromosome Location
Chr.3: 150801289 - 150802935 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199500.2 NP_955794.1 3 - 4 383 121
GenBank mRNA AY206668.1 - 3 - 4 331 121
BC086389.1 - 3 - 4 344 121
FM036735.1 - 2 - 3 168 121
FM046244.1 - 3 - 4 351 121

View Full Product Details