Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndrg4
Entrez Gene ID
64457
Gene Name
NDRG family member 4
Gene Aliases
Bdm1, Ndr4, smap8
Chromosome Location
Chr.19: 9766439 - 9802387 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271091.1 NP_001258020.1 13 - 14 1043 64
NM_001271092.1 NP_001258021.1 13 - 14 1043 64
NM_001271093.1 NP_001258022.1 13 - 14 990 64
NM_001271094.1 NP_001258023.1 13 - 14 990 64
NM_001271095.1 NP_001258024.1 11 - 12 908 64
NM_031967.2 NP_114173.2 11 - 12 908 64
XM_006255123.2 XP_006255185.1 13 - 14 985 64
GenBank mRNA AF045564.1 - 11 - 12 840 64
AY217029.1 - 11 - 12 879 64
AY217030.1 - 13 - 14 990 64
AY217031.1 - 13 - 14 990 64
AY217032.1 - 13 - 14 1038 64
AY217033.1 - 13 - 14 1038 64
AY255791.1 - 11 - 12 879 64
CB585874.1 - 7 - 8 465 64
CB713901.1 - 4 - 5 223 64

View Full Product Details