The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039004.1 | NP_001034093.1 | 5 - 6 | 633 | 98 |
XM_006255132.2 | XP_006255194.1 | 5 - 6 | 1266 | 98 | |
GenBank mRNA | BC098748.1 | - | 5 - 6 | 633 | 98 |
BC168149.1 | - | 5 - 6 | 1272 | 98 | |
FQ229498.1 | - | 5 - 6 | 674 | 98 |