The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139111.1 | NP_620811.1 | 3 - 4 | 495 | 88 |
GenBank mRNA | AF253064.2 | - | 2 - 3 | 336 | 88 |
AF253065.2 | - | 3 - 4 | 495 | 88 |