Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tk2
Entrez Gene ID
291824
Gene Name
thymidine kinase 2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.19: 917203 - 939221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106166.1 NP_001099636.1 1 - 2 145 152

View Full Product Details