Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Setd6
Entrez Gene ID
291844
Gene Name
SET domain containing 6
Gene Aliases
RGD1560538
Chromosome Location
Chr.19: 9758613 - 9766362 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106167.1 NP_001099637.1 6 - 7 1144 87

View Full Product Details