Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dok4
Entrez Gene ID
361364
Gene Name
docking protein 4
Gene Aliases
-
Chromosome Location
Chr.19: 10563390 - 10575109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108438.2 NP_001101908.1 7 - 8 1031 76
XM_008772317.1 XP_008770539.1 6 - 7 971 76
GenBank mRNA BC158749.1 - 7 - 8 1031 76

View Full Product Details