The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012040.1 | NP_001012040.1 | 5 - 6 | 1164 | 82 |
XM_006251640.1 | XP_006251702.1 | 5 - 6 | 1202 | 82 | |
GenBank mRNA | BC082100.1 | - | 5 - 6 | 1164 | 82 |
FQ219794.1 | - | 5 - 6 | 1202 | 82 |