Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nid2
Entrez Gene ID
302248
Gene Name
nidogen 2 (osteonidogen)
Gene Aliases
NID-2
Chromosome Location
Chr.15: 4893999 - 4951893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012005.2 NP_001012005.2 17 - 18 3807 81
GenBank mRNA BC088325.1 - 1 - 2 405 81
BC169012.1 - 17 - 18 3807 81

View Full Product Details