Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcnma1
- Entrez Gene ID
-
83731
- Gene Name
- potassium channel, calcium activated large conductance subfamily M alpha, member 1
- Gene Aliases
-
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
- Chromosome Location
-
Chr.15: 344204 - 1048849 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB248959.1 |
- |
4 - 5 |
204 |
81 |
View Full Product Details