Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fut11
Entrez Gene ID
286971
Gene Name
fucosyltransferase 11 (alpha (1,3) fucosyltransferase)
Gene Aliases
-
Chromosome Location
Chr.15: 4031504 - 4035064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173308.2 NP_775430.2 1 - 2 729 112
GenBank mRNA AJ535753.1 - 1 - 2 754 112
BC079316.1 - 1 - 2 729 112

View Full Product Details