Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clvs2
Entrez Gene ID
361459
Gene Name
clavesin 2
Gene Aliases
Clavesin-2, RGD1306801, Rlbp1l2
Chromosome Location
Chr.1: 25173089 - 25174975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108459.1 NP_001101929.1 - 995 124

View Full Product Details