Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkd1
Entrez Gene ID
364952
Gene Name
naked cuticle homolog 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.19: 19434731 - 19509087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271381.1 NP_001258310.1 8 - 9 706 112
XM_008772413.1 XP_008770635.1 7 - 8 871 112
GenBank mRNA BC168970.1 - 7 - 8 1068 112

View Full Product Details