Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irx5
Entrez Gene ID
498918
Gene Name
iroquois homeobox 5
Gene Aliases
-
Chromosome Location
Chr.19: 16415850 - 16420614 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030044.1 NP_001025215.1 1 - 2 249 98
GenBank mRNA DQ109812.1 - 1 - 2 249 98

View Full Product Details