Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mylk3
Entrez Gene ID
291926
Gene Name
myosin light chain kinase 3
Gene Aliases
MLCK, RGD1305801
Chromosome Location
Chr.19: 27388163 - 27442451 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255355.2 XP_006255417.1 1 - 2 189 61

View Full Product Details