Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3galnt2
Entrez Gene ID
291212
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
RGD1306946
Chromosome Location
Chr.17: 54027859 - 54070399 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144851.1 NP_001138323.1 2 - 3 596 124
XM_006254007.2 XP_006254069.1 2 - 3 533 124
XM_008771701.1 XP_008769923.1 2 - 3 532 124
XM_008771702.1 XP_008769924.1 2 - 3 532 124

View Full Product Details