Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtpap
Entrez Gene ID
307050
Gene Name
mitochondrial poly(A) polymerase
Gene Aliases
Papd1
Chromosome Location
Chr.17: 56046507 - 56068209 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107359.1 NP_001100829.1 3 - 4 568 109
XM_006254071.2 XP_006254133.1 3 - 4 671 109

View Full Product Details