The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017030.2 | NP_058726.2 | 14 - 15 | 1514 | 90 |
GenBank mRNA | BC078855.1 | - | 14 - 15 | 1514 | 90 |
M14634.1 | - | 14 - 15 | 1515 | 90 |