The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020543.4 | NP_065418.3 | 1 - 2 | 1032 | 68 |
GenBank mRNA | AF218846.1 | - | 1 - 2 | 1032 | 68 |