Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc20a1
Entrez Gene ID
81826
Gene Name
solute carrier family 20 (phosphate transporter), member 1
Gene Aliases
PiT-1
Chromosome Location
Chr.3: 121725399 - 121739160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031148.1 NP_112410.1 5 - 6 841 86
XM_006234991.2 XP_006235053.1 5 - 6 1065 86
GenBank mRNA AB000489.1 - 5 - 6 881 86

View Full Product Details