Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndrg4
Entrez Gene ID
64457
Gene Name
NDRG family member 4
Gene Aliases
Bdm1, Ndr4, smap8
Chromosome Location
Chr.19: 9766439 - 9802387 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271091.1 NP_001258020.1 5 - 6 515 110
NM_001271092.1 NP_001258021.1 5 - 6 515 110
NM_001271093.1 NP_001258022.1 5 - 6 462 110
NM_001271094.1 NP_001258023.1 5 - 6 462 110
NM_001271095.1 NP_001258024.1 3 - 4 380 110
NM_031967.2 NP_114173.2 3 - 4 380 110
XM_006255123.2 XP_006255185.1 5 - 6 457 110
GenBank mRNA AF045564.1 - 3 - 4 312 110
AY217029.1 - 3 - 4 351 110
AY217030.1 - 5 - 6 462 110
AY217031.1 - 5 - 6 462 110
AY217032.1 - 5 - 6 510 110
AY217033.1 - 5 - 6 510 110
AY255791.1 - 3 - 4 351 110
FM054107.1 - 3 - 4 333 110

View Full Product Details