The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053613.1 | NP_446065.1 | 2 - 2 | 1054 | 63 |
XM_008768839.1 | XP_008767061.1 | 2 - 2 | 63 | ||
GenBank mRNA | AF462390.1 | - | 2 - 2 | 63 | |
AY028438.1 | - | 2 - 2 | 1054 | 63 |