Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim63
Entrez Gene ID
140939
Gene Name
tripartite motif containing 63, E3 ubiquitin protein ligase
Gene Aliases
Murf, Murf1, Rnf28
Chromosome Location
Chr.5: 152533362 - 152547138 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080903.1 NP_543179.1 7 - 8 1079 78
GenBank mRNA AY059627.1 - 7 - 8 1079 78
BC061824.1 - 7 - 8 1066 78

View Full Product Details