Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
171409
Gene Name
troponin T type 1 (skeletal, slow)
Gene Aliases
Fang2, sTnT, tnTs
Chromosome Location
Chr.1: 72889270 - 72899629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277260.1 NP_001264189.1 12 - 13 749 90
NM_001277261.2 NP_001264190.1 11 - 12 716 90
NM_001277262.2 NP_001264191.1 11 - 12 719 90
NM_134388.2 NP_599215.2 12 - 13 752 90
GenBank mRNA AF399874.1 - 11 - 12 90
AY334079.1 - 11 - 12 696 90
AY334080.1 - 11 - 12 693 90
AY334081.1 - 10 - 11 663 90
AY334082.1 - 10 - 11 660 90
FQ215948.1 - 10 - 11 749 90

View Full Product Details