Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3rf1
Entrez Gene ID
306417
Gene Name
SH3 domain containing ring finger 1
Gene Aliases
Posh, Sh3md2
Chromosome Location
Chr.16: 32062339 - 32225777 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198764.1 NP_942059.1 3 - 4 754 92
XM_008771225.1 XP_008769447.1 2 - 3 871 92
GenBank mRNA AF515735.1 - 3 - 4 754 92

View Full Product Details