Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpped2
Entrez Gene ID
362185
Gene Name
metallophosphoesterase domain containing 2
Gene Aliases
C11orf8, C11orf8h
Chromosome Location
Chr.3: 97720308 - 97899735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198778.2 NP_942073.2 4 - 5 694 138
XM_006234680.2 XP_006234742.1 4 - 5 2069 138
XM_006234681.2 XP_006234743.1 4 - 5 771 138
XM_006234682.2 XP_006234744.1 4 - 5 858 138
XM_006234683.2 XP_006234745.1 4 - 5 773 138
XM_008762104.1 XP_008760326.1 4 - 5 1493 138
XM_008762105.1 XP_008760327.1 3 - 4 1753 138
GenBank mRNA AY093422.1 - 4 - 5 744 138
BC161828.1 - 4 - 5 776 138
EV768250.1 - 2 - 3 329 138

View Full Product Details