The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199103.1 | NP_954534.1 | 3 - 4 | 521 | 64 |
XM_006229580.2 | XP_006229642.1 | 5 - 6 | 1769 | 64 | |
XM_006229584.2 | XP_006229646.1 | 5 - 6 | 755 | 64 | |
XM_006229585.2 | XP_006229647.1 | 4 - 5 | 715 | 64 | |
XM_008773600.1 | XP_008771822.1 | 4 - 5 | 4829 | 64 | |
XM_008773601.1 | XP_008771823.1 | 5 - 6 | 1772 | 64 | |
XM_008773602.1 | XP_008771824.1 | 5 - 6 | 1786 | 64 | |
XM_008773603.1 | XP_008771825.1 | 5 - 6 | 1777 | 64 | |
XM_008773604.1 | XP_008771826.1 | 4 - 5 | 4826 | 64 | |
XM_008773605.1 | XP_008771827.1 | 4 - 5 | 4826 | 64 | |
XM_008773606.1 | XP_008771828.1 | 4 - 5 | 4823 | 64 | |
XM_008773607.1 | XP_008771829.1 | 5 - 6 | 4869 | 64 | |
XM_008773608.1 | XP_008771830.1 | 5 - 6 | 759 | 64 | |
XM_008773609.1 | XP_008771831.1 | 5 - 6 | 967 | 64 | |
XM_008773610.1 | XP_008771832.1 | 4 - 5 | 719 | 64 | |
GenBank mRNA | AY392762.1 | - | 3 - 4 | 521 | 64 |