The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201271.1 | NP_958428.1 | 5 - 6 | 878 | 72 |
GenBank mRNA | AY299399.1 | - | 5 - 6 | 878 | 72 |