Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctrb1
Entrez Gene ID
24291
Gene Name
chymotrypsinogen B1
Gene Aliases
Ctrb
Chromosome Location
Chr.19: 43906344 - 43911020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012536.1 NP_036668.1 2 - 3 161 106
GenBank mRNA FQ232114.1 - 2 - 3 106
FQ233595.1 - 2 - 3 106
FQ234337.1 - 2 - 3 106
FQ234339.1 - 2 - 3 106

View Full Product Details