Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psap
Entrez Gene ID
25524
Gene Name
prosaposin
Gene Aliases
SGP-1, SGP1A
Chromosome Location
Chr.20: 29831302 - 29856876 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190236.1 NP_001177165.1 1 - 2 121 97
NM_001190237.1 NP_001177166.1 1 - 2 121 97
NM_001190238.1 NP_001177167.1 1 - 2 121 97
NM_013013.2 NP_037145.2 1 - 2 121 97
XM_006256403.1 XP_006256465.1 1 - 2 133 97
GenBank mRNA BC061759.1 - 1 - 2 55 97
CK365248.1 - 1 - 2 121 97
FQ217536.1 - 1 - 2 72 97
M19936.1 - 1 - 2 71 97
S81353.1 - 1 - 2 71 97

View Full Product Details