The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013033.2 | NP_037165.1 | 10 - 11 | 1296 | 64 |
GenBank mRNA | AB000729.1 | - | 10 - 11 | 1322 | 64 |
BC081827.1 | - | 10 - 11 | 1328 | 64 | |
D16101.1 | - | 10 - 11 | 1296 | 64 | |
U03120.1 | - | 10 - 11 | 1254 | 64 |