The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022681.2 | NP_073172.2 | 4 - 4 | 3067 | 101 |
GenBank mRNA | AF234680.1 | - | 1 - 1 | 2087 | 101 |