Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mvd
Entrez Gene ID
81726
Gene Name
mevalonate (diphospho) decarboxylase
Gene Aliases
Mvpd
Chromosome Location
Chr.19: 55258521 - 55269108 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031062.2 NP_112324.1 3 - 4 298 153
GenBank mRNA BC081784.1 - 3 - 4 283 153
U53706.1 - 3 - 4 299 153

View Full Product Details